missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzamide, 99%

Product Code. 11342917
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342917 25 g 25g
11322917 1 g 1g
11332917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342917 Supplier Thermo Scientific Alfa Aesar Supplier No. L16176.14
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 170229-98-8 | C8H8BrNO | 214.062 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170229-98-8
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.062
MDL Number MFCD00672914
InChI Key ZPFPOZMNEKPBIF-UHFFFAOYSA-N
Synonym benzamide,4-bromo-3-methyl, pubchem3804, 4-bromo-3-methyl-benzamide, acmc-1c5o5, 4-bromanyl-3-methyl-benzamide
PubChem CID 7020869
IUPAC Name 4-bromo-3-methylbenzamide
SMILES CC1=C(C=CC(=C1)C(=O)N)Br

Specifications

Melting Point 177°C to 182°C
Quantity 25 g
Beilstein 868074
Formula Weight 214.06
Percent Purity 99%
Chemical Name or Material 4-Bromo-3-methylbenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.