missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzonitrile, 98%

Product Code. 11457056
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457056 25 g 25g
11309419 1 g 1g
11447056 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457056 Supplier Thermo Scientific Alfa Aesar Supplier No. B25377.14
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 41963-20-6 | C8H6BrN | 196.047 g/mol

4-bromo-3-methylbenzenecarbonitrile is a nitrile compound for proteomics research & a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-bromo-3-methylbenzenecarbonitrile is a nitrile compound for proteomics research & a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in refrigerator at (2-4°C). Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41963-20-6
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00031538
InChI Key SKXUZFJOLNNWIG-UHFFFAOYSA-N
Synonym 3-methyl-4-bromobenzonitrile, 4-bromo-3-methyl-benzonitrile, 4-bromo-3-methylbenzenecarbonitrile, 4-bromo-3-methylbenzonitrile, 2-bromo-5-cyanotoluene, pubchem3808, 4-bromo-m-tolunitrile, acmc-209jmj, ksc497m9n, 4-bromo-3-methyl benzonitrile
PubChem CID 2737494
IUPAC Name 4-bromo-3-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)C#N)Br

Specifications

Melting Point 52°C to 54°C
Boiling Point 129°C to 130°C (5 mmHg)
Flash Point 96°C (205°F)
Quantity 25 g
UN Number UN3439
Beilstein 2354972
Solubility Information Insoluble in water.
Formula Weight 196.05
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-methylbenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.