missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoic hydrazide 98%, ACROS Organics™

Product Code. 10085503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10085503 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10085503 Supplier Thermo Scientific Acros Supplier No. 386460050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5933-32-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 166.0°C to 168.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H7BrN2O
Linear Formula BrC6H4C(O)NHNH2
MDL Number MFCD00007602
Quantity 5 g
Synonym 4-bromobenzhydrazide, p-bromobenzhydrazide, 4-bromobenzoic hydrazide, p-bromobenzohydrazide, p-bromobenzoic acid hydrazide, 4-bromobenzoylhydrazine, 4-bromobenzoic acid hydrazide, benzoic acid, 4-bromo-, hydrazide, 4-bromo-benzoic acid hydrazide, p-bromobenzoylhydrazine
InChI Key UYIMBYKIIMYFPS-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C(=O)NN)Br
IUPAC Name 4-bromobenzohydrazide
Molecular Weight (g/mol) 215.05
PubChem CID 22219
Formula Weight 215.05
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Bromobenzoic hydrazide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzoic hydrazide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.