missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethylphenylboronic acid, TRC

Product Code. 30342598
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342598 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342598 Supplier Toronto Research Chemicals Supplier No. TRCB694828250MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromomethylphenylboronic acid
CAS 68162-47-0
Molecular Formula C7H8BBrO2
Quantity 250 mg
InChI Formula InChI=1S/C7H8BBrO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4,10-11H,5H2
SMILES OB(O)c1ccc(CBr)cc1
IUPAC Name [4-(bromomethyl)phenyl]boronic acid
Molecular Weight (g/mol) 214.85
Formula Weight 213.9801
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.