missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenoxyacetic acid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318777
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318777 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318777 Supplier Dr. Ehrenstorfer Supplier No. DREA10737000AL100
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromophenoxyacetic Acid
CAS 1878-91-7
Analyte or Component Names 4-Bromophenoxyacetic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H7 Br O3
Certifications/Compliance ISO 17034
Synonym 4-Bromophenoxyacetic Acid, Acetic acid, (4-bromophenoxy)- (9CI), Acetic acid, (p-bromophenoxy)- (6CI,7CI,8CI), 2-(4-Bromophenoxy)acetic acid, (4-Bromophenoxy)acetic acid, NSC 65085, p-Bromophenoxyacetic acid, Acetic acid, 2-(4-bromophenoxy)-
InChI Formula InChI=1S/C8H7BrO3/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4H,5H2,(H,10,11)
SMILES OC(=O)COc1ccc(Br)cc1
IUPAC Name 2-(4-bromophenoxy)acetic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 231.0434
Quantity 1 mL
Formula Weight 229.9579 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.