missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxybenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10364962
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10364962 10 g 10g
10601123 1 g 1g
10640856 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10364962 Supplier Thermo Scientific Maybridge Supplier No. TL00441EA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1138-56-3
Molecular Formula C10H13ClO3S
Molecular Weight (g/mol) 248.721
MDL Number MFCD00052344
InChI Key HGKWMUBXVMFXNC-UHFFFAOYSA-N
Synonym 4-n-butoxy benzenesulfonyl chloride, 4-butoxybenzene-1-sulfonyl chloride, 4-n-butoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-butoxy, 4-butoxy-benzenesulfonyl chloride, 4-n-butoxybenzenesulfonyl chloride, 4-butoxybenzenesulfonylchloride, pubchem5072, 4-n-butoxy benzenes, buttpark 9650-54
PubChem CID 517990
IUPAC Name 4-butoxybenzenesulfonyl chloride
SMILES CCCCOC1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)3OC6H4SO2Cl
Quantity 10 g
Formula Weight 248.73
Percent Purity 97%
Chemical Name or Material 4-Butoxybenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.