missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-(4-fluorophenyl)butan-1-one, 97%, BLD Pharm™

Product Code. 30981038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30981038 500 g 500g
30977685 100 g 100g
30977757 10 g 10g
30970670 5 g -
30973874 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30981038 Supplier BLD Pharm Supplier No. BD0032500g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3874-54-2
Molecular Formula C10H10ClFO
MDL Number MFCD00001007
Quantity 500 g
SMILES O=C(C1=CC=C(F)C=C1)CCCCl
Molecular Weight (g/mol) 200.64
Percent Purity 97%
Chemical Name or Material 4-Chloro-1-(4-fluorophenyl)butan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.