Learn More
4-Chloro-2-fluoro-1-iodobenzene, 98%
Product Code. 11375259
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375259 5 g 5g
11385259 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11375259 Supplier Thermo Scientific Alfa Aesar Supplier No. A19376.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
CAS: 6797-79-1 | C6H3ClFI | 256.44 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6797-79-1
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.44
MDL Number MFCD00079729
InChI Key RSTFBOIFYXJIMR-UHFFFAOYSA-N
Synonym 4-chloro-2-fluoroiodobenzene, 2-fluoro-4-chloroiodobenzene, 1-chloro-3-fluoro-4-iodobenzene, 4-chloro-2-fluoro-1-iodo-benzene, 4-chloro-2-fluoro-iodobenzene, pubchem3435, acmc-1b71k, 2-iodo-5-chlorofluorobenzene, 2-iodo-5-chlorofluoro benzene, 3-fluoro-4-iodophenyl chloride
PubChem CID 2736562
IUPAC Name 4-chloro-2-fluoro-1-iodobenzene
SMILES FC1=C(I)C=CC(Cl)=C1
Specifications
Density 1.98
Boiling Point 217°C
Flash Point >110°C (230°F)
Refractive Index 1.606
Quantity 5 g
Beilstein 6288419
Sensitivity Light sensitive
Formula Weight 256.45
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoro-1-iodobenzene