missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoroaniline, 98%

Product Code. 11413177
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413177 100 g 100g
11403177 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413177 Supplier Thermo Scientific Alfa Aesar Supplier No. A16131.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 57946-56-2 | C6H5ClFN | 145.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57946-56-2
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00010625
InChI Key CSFDTBRRIBJILD-UHFFFAOYSA-N
Synonym 2-fluoro-4-chloroaniline, 4-chloro-2-fluorobenzenamine, 4-chloro-2-fluoro-phenylamine, benzenamine, 4-chloro-2-fluoro, 4-chloro-2-fluoro-aniline, 4-chloro-2-fluorophenylamine, pubchem1514, 4-chloro-2fluoroaniline, 4chloro-2-fluoroaniline, acmc-1at8a
PubChem CID 93898
IUPAC Name 4-chloro-2-fluoroaniline
SMILES NC1=CC=C(Cl)C=C1F

Specifications

Density 1.311
Quantity 100 g
Refractive Index 1.56
UN Number UN2810
Beilstein 1934804
Boiling Point 92°C (2 mmHg)
Formula Weight 145.56
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.