missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzaldehyde, 98%

Product Code. 11413147
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413147 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413147 Supplier Thermo Scientific Alfa Aesar Supplier No. A16110.09
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61072-56-8 | C7H4ClFO | 158.556 g/mol

4-Chloro-2-fluorobenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-2-fluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 61072-56-8
Melting Point 58°C to 60°C
Molecular Formula C7H4ClFO
MDL Number MFCD00143282
Quantity 10 g
Beilstein 3537705
Sensitivity Air Sensitive
Synonym 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985
Solubility Information Insoluble in water.
InChI Key UVGYSEIWAOOIJR-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)F)C=O
IUPAC Name 4-chloro-2-fluorobenzaldehyde
Molecular Weight (g/mol) 158.556
PubChem CID 2724908
Formula Weight 158.56
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.