missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorophenol, 98+%, Thermo Scientific™

Product Code. 10491543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10491543 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10491543 Supplier Thermo Scientific Acros Supplier No. 378210050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-62-9
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.55
MDL Number MFCD00075030
InChI Key ZKMUKBBWORLNLA-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorophenol, phenol, 4-chloro-2-fluoro, 4-chloro-2-fluoro-phenol, 4-chloro-fluorophenol, pubchem2861, acmc-1ct1y, ksc222g4r, 4-chloro-2-fluorophenol, attercop-chm at106713
PubChem CID 181975
IUPAC Name 4-chloro-2-fluorophenol
SMILES OC1=CC=C(Cl)C=C1F

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 20°C
Color Colorless to Pink
Density 1.3780g/mL
Boiling Point 103°C to 104°C (50.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3FOH
Refractive Index 1.5330 to 1.535
Quantity 5 g
Specific Gravity 1.378
Solubility Information Solubility in water: insoluble
Formula Weight 146.55
Percent Purity 98+%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Chloro-2-fluorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.