missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-iodoanisole, 98%, Thermo Scientific™

Product Code. 11441488
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441488 25 g 25g
11431488 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441488 Supplier Thermo Scientific Alfa Aesar Supplier No. A13840.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
An independent synthesis of 4-chloro-2-iodoanisole has shown that this is chlorinating agents, brominating agents and iodinating agents. 4-Chloro-2-iodoanisole (2) was demethylated and subsequently alkylated with 4-chloro-2-fluorobenzyl bromide, an aryl Grignard reagent was formed with it.

Solubility
Soluble in methanol.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52807-27-9
Molecular Formula C7H6ClIO
Molecular Weight (g/mol) 268.48
MDL Number MFCD00052858
InChI Key CXXXAGIYCFOGHA-UHFFFAOYSA-N
Synonym 4-chloro-2-iodoanisole, 5-chloro-2-methoxyiodobenzene, 4-chloro-2-iodo-1-methoxy-benzene, 4-chloroiodoanisole, pubchem14310, 2-iodo-4-chloroanisole, acmc-209l1x, 4-chloro-2-iodo-1-methoxybenzen, benzene,4-chloro-2-iodo-1-methoxy, 4-chloro-2-iodoanisol;chloro-2-iodoanisole
PubChem CID 2800978
IUPAC Name 4-chloro-2-iodo-1-methoxybenzene
SMILES COC1=CC=C(Cl)C=C1I

Specifications

Melting Point 44°C to 48°C
Boiling Point 133°C to 135°C (15 mmHg)
Quantity 25 g
Sensitivity Light sensitive
Solubility Information Soluble in methanol.
Formula Weight 268.48
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-iodoanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.