missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxy-5-methylaniline, 85%, Tech., Thermo Scientific™

Product Code. 10400052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10400052 50 g 50g
10555101 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10400052 Supplier Thermo Scientific Acros Supplier No. 331360500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6376-14-3
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.62
MDL Number MFCD00007847
InChI Key XBAPOWUMJRIKAV-UHFFFAOYSA-N
Synonym chlorocresidine, 4-chloro-5-methyl-o-anisidine, o-anisidine, 4-chloro-5-methyl, 4-chloro-2-methoxy-5-methylphenylamine, 4-chloro-2-methoxy-5-methylanilin, 2-methoxy-5-methyl-4-chloroaniline, 4-chloro-2-methoxy-5-methyl-aniline, 4-chloro-2-methoxy-5-methyl-phenylamine, benzenamine,4-chloro-2-methoxy-5-methyl, tech.,
PubChem CID 80782
IUPAC Name 4-chloro-2-methoxy-5-methylaniline
SMILES COC1=CC(Cl)=C(C)C=C1N

Specifications

CAS Min % 80.0
CAS Max % 100.0
Melting Point 97°C to 103°C
Color Black
Infrared Spectrum Authentic
Assay Percent Range 85%
Packaging Glass bottle
Quantity 50 g
Formula Weight 171.63
Percent Purity ≥80%
Grade Technical
Physical Form Crystalline Solid
Chemical Name or Material 4-Chloro-2-methoxy-5-methylaniline, tech., 85%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.