missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylaniline Hydrochloride, TRC

Product Code. 30135322
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135322 2.5 g 2.5g
18002010 500 mg 500mg
18012720 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135322 Supplier Toronto Research Chemicals Supplier No. TRCC3744802.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3165-93-3
Molecular Formula C7H8ClN • HCl
Molecular Weight (g/mol) 141.603646
Synonym Benzenamine, 4-chloro-2-methyl-, hydrochloride (9CI), o-Toluidine, 4-chloro-, hydrochloride (8CI), 2-Methyl-4-chloroaniline hydrochloride, 4-Chloro-2-methylaniline hydrochloride, 4-Chloro-2-methylbenzenamine hydrochloride, 4-Chloro-o-toluidine hydrochloride, Azogene Fast Red TR, C.I. 37085, Devol Red K, Devol Red TR, Neutrosel Red TRVA, p-Chloro-o-toluidine hydrochloride, 4-Chloro-2-methylaniline Hydrochloride
IUPAC Name 4-chloro-2-methylaniline;hydrochloride
SMILES CC1=C(C=CC(=C1)Cl)N.Cl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H8ClN.ClH/c1-5-4-6(8)2-3-7(5)9;/h2-4H,9H2,1H3;1H
Formula Weight 177.0112
Percent Purity >95%
Chemical Name or Material 4-Chloro-2-methylaniline Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.