missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dimethylphenoxy)acetic Acid, TRC

Product Code. 30180255
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180255 25 mg 25mg
30179734 100 mg 100mg
30179945 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180255 Supplier Toronto Research Chemicals Supplier No. TRCC36748525MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19545-95-0
Molecular Formula C10H11ClO3
Molecular Weight (g/mol) 214.65
Synonym (4-Chloro-3,5-dimethylphenoxy)acetic Acid, [(4-chloro-3,5-xylyl)oxy]acetic Acid;2-(4-Chloro-3,5-dimethylphenoxy)acetic Acid;
IUPAC Name 2-(4-chloro-3,5-dimethylphenoxy)acetic acid
SMILES ClC1=C(C)C=C(OCC(O)=O)C=C1C

Tekniske data

Quantity 25 mg
InChI Formula InChI=1S/C10H11ClO3/c1-6-3-8(14-5-9(12)13)4-7(2)10(6)11/h3-4H,5H2,1-2H3,(H,12,13)
Formula Weight 214.4
Chemical Name or Material 4-(Chloro-3,5-dimethylphenoxy)acetic Acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.