missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-4'-fluorobutyrophenone, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-4'-fluorobutyrophenone |
| CAS | 3874-54-2 |
| Molecular Formula | C10 H10 Cl F O |
| Quantity | 2.5 g |
| Synonym | 4-Chloro-4'-fluorobutyrophenone, 1-Butanone, 4-chloro-1-(4-fluorophenyl)- (9CI, ACI), 4-Chloro-1-(4-fluorophenyl)-1-butanone (ACI), Butyrophenone, 4-chloro-4'-fluoro- (6CI, 7CI, 8CI), 1-Chloro-4-(4-fluorophenyl)-4-oxobutane, 3-(4-Fluorobenzoyl)propyl chloride, 3-(4-Fluorophenylcarbonyl)-1-chloropropane, 3-(p-Fluorobenzoyl)propyl chloride, 3-Chloropropyl 4-fluorophenyl ketone, 4-(4-Fluorophenyl)-4-oxobutyl chloride, 4-(p-Fluorophenyl)-4-oxobutyl chloride, 4-Chloro-1-(p-fluorophenyl)-1-butanone, 4-Chloro-1-(p-fluorophenyl)-1-oxobutane, 4-Chloro-4'-chlorobutyrophenone, 4-Chloro-4'-fluorobutyrophenone, 4-Chloro-p-fluorobutyrophenone, 4-p-Fluorophenyl-4-oxo-1-chlorobutane, 4'-Fluoro-4-chlorobutyrophenone, NSC 87082, p-Fluoro-4-chlorobutyrophenone, p-Fluoro-γ-chlorobutyrophenone, p-Fluoro-ω-chlorobutyrophenone, γ-Chloro-4-fluorobutyrophenone, γ-Chloro-p-fluorobutyrophenone, ω-Chloro-4-fluorobutyrophenone, ω-Chloro-p-fluorobutyrophenone, ω-Chloropropyl 4-fluorophenyl ketone |
| InChI Formula | InChI=1S/C10H10ClFO/c11-7-1-2-10(13)8-3-5-9(12)6-4-8/h3-6H,1-2,7H2 |
| SMILES | Fc1ccc(cc1)C(=O)CCCCl |
| IUPAC Name | 4-chloro-1-(4-fluorophenyl)butan-1-one |
| Molecular Weight (g/mol) | 200.64 |
| Formula Weight | 200.0404 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction