missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-6-fluoroquinoline

Product Code. 11369999
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369999 1 g 1g
11359999 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369999 Supplier Alfa Aesar™ Supplier No. H50540.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 391-77-5
Molecular Formula C9H5ClFN
Molecular Weight (g/mol) 181.59
MDL Number MFCD00278783
InChI Key CKTQPWIDUQGUGG-UHFFFAOYSA-N
Synonym 4-chloro-6-fluoro-quinoline, quinoline, 4-chloro-6-fluoro, pubchem5845, 6-fluoro-4-chloroquinoline, ksc493m4t, 4-chloranyl-6-fluoranyl-quinoline, 4-chloro-6-fluoroquinoline
PubChem CID 2736586
IUPAC Name 4-chloro-6-fluoroquinoline
SMILES FC1=CC=C2N=CC=C(Cl)C2=C1

Specifications

Quantity 1 g
Formula Weight 181.6
Chemical Name or Material 4-Chloro-6-fluoroquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.