missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-methylpyrimidine, 97%

Product Code. 15421969
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421969 1 g 1g
15441969 250 mg 250mg
15431969 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421969 Supplier Thermo Scientific Alfa Aesar Supplier No. H64667.03
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3435-25-4 | C5H5ClN2 | 128.559 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3435-25-4
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD02322991
InChI Key MVAXKFAQKTWRAH-UHFFFAOYSA-N
Synonym 4-methyl-6-chloro pyrimidine, 4-methyl-6-chloropyrimidine, pyrimidine, 4-chloro-6-methyl, 4-chloro-6-methyl-pyrimidine, 6-chloro-4-methylpyrimidine, 4-chloro-6-methyl-1,3-diazine, pubchem13240, 6-methyl-4-chloropyrimidin, ksc269g3h, pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci
PubChem CID 581796
IUPAC Name 4-chloro-6-methylpyrimidine
SMILES CC1=CC(=NC=N1)Cl

Specifications

Color Yellow
Quantity 1 g
Beilstein 108656
Solubility Information Slightly soluble in water.
Formula Weight 128.56
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-6-methylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.