Learn More
4-Chloro-DL-phenylalanine, 98+%
Product Code. p-7021120
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11400933
Brand: Thermo Scientific Alfa Aesar A13323.09
Request Bulk or Custom Format
Edge
CAS: 7424-00-2 | C9H10ClNO2 | 199.63 g/mol
4-Chloro-DL-phenylalanine is an irreversible tryptophan hydroxylase inhibitor that depletes serotonin in the brain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloro-DL-phenylalanine is an irreversible tryptophan hydroxylase inhibitor that depletes serotonin in the brain.
Solubility
Soluble to 10 mM in 1eq. NaOH and to 5 mM in water with gentle warming.
Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7424-00-2
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00002601
InChI Key NIGWMJHCCYYCSF-UHFFFAOYNA-N
Synonym fenclonine, 4-chloro-dl-phenylalanine, p-chlorophenylalanine, 2-amino-3-4-chlorophenyl propanoic acid, 4-chlorophenylalanine, dl-4-chlorophenylalanine, dl-p-chlorophenylalanine, fenchlonine, fenclonin, phenylalanine, 4-chloro
PubChem CID 4652
IUPAC Name 2-amino-3-(4-chlorophenyl)propanoic acid
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O
Specifications
Melting Point ∼265°C (decomposition)
Beilstein 2805758
Solubility Information Soluble to 10 mM in 1eq. NaOH and to 5 mM in water with gentle warming.
Quantity 10 g
Formula Weight 199.64
Percent Purity ≥98%
Chemical Name or Material 4-Chloro-DL-phenylalanine