Learn More
4-Chloro Perphenazine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloro Perphenazine |
| CAS | 875256-24-9 |
| Molecular Formula | C21H26ClN3OS |
| Quantity | 5 mg |
| Synonym | 4-[3-[4-Chlorophenothiazin-10-yl]propyl]-1-piperazineethanol, 4-[3-(4-Chloro-10H-phenothiazin-10-yl)propyl]-1-piperazineethanol |
| InChI Formula | InChI=1S/C21H26ClN3OS/c22-17-5-3-7-19-21(17)27-20-8-2-1-6-18(20)25(19)10-4-9-23-11-13-24(14-12-23)15-16-26/h1-3,5-8,26H,4,9-16H2 |
| SMILES | OCCN1CCN(CCCN2c3ccccc3Sc4c(Cl)cccc24)CC1 |
| IUPAC Name | 2-[4-[3-(4-chlorophenothiazin-10-yl)propyl]piperazin-1-yl]ethanol |
| Molecular Weight (g/mol) | 403.97 |