missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyronitrile, 97%

Product Code. 11395239
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395239 50 g 50g
11305249 250 g 250g
11385239 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11395239 Supplier Thermo Scientific Alfa Aesar Supplier No. A18942.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 628-20-6 | C4H6ClN | 103.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-20-6
Molecular Formula C4H6ClN
Molecular Weight (g/mol) 103.55
MDL Number MFCD00001972
InChI Key ZFCFBWSVQWGOJJ-UHFFFAOYSA-N
Synonym 4-chlorobutyronitrile, butanenitrile, 4-chloro, butyronitrile, 4-chloro, gamma-chlorobutyronitrile, 1-chloro-3-cyanopropane, 4-chloro-butyronitrile, .gamma.-chlorobutyronitrile, 4chlorobutyronitrile, 4-chlorobutyroonitrile, 4-chloro-butanenitrile
PubChem CID 12336
IUPAC Name 4-chlorobutanenitrile
SMILES ClCCCC#N

Specifications

Density 1.084
Boiling Point 195°C to 197°C
Flash Point 85°C (185°F)
Linear Formula Cl(CH2)3CN
Refractive Index 1.444
Quantity 50 g
UN Number UN3276
Beilstein 1633582
Formula Weight 103.55
Percent Purity 97%
Chemical Name or Material 4-Chlorobutyronitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.