missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromandelic acid, 98%

Product Code. 11471317
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471317 5 g 5g
11481317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471317 Supplier Thermo Scientific Alfa Aesar Supplier No. B24264.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 492-86-4 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-86-4
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00042724
InChI Key BWSFWXSSALIZAU-UHFFFAOYSA-N
Synonym 4-chloromandelic acid, 2-4-chlorophenyl-2-hydroxyacetic acid, p-chloromandelic acid, p-chloro mandelic acid, 4-chloro-dl-mandelic acid, 4-chloromandelic acid, pract., benzeneacetic acid, 4-chloro-.alpha.-hydroxy, dl-4-chloromandelic acid, mandelic acid, p-chloro, 4-chlorophenyl hydroxy acetic acid
PubChem CID 10299
IUPAC Name 2-(4-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)O)Cl

Specifications

Melting Point 117°C to 122°C
Quantity 5 g
Beilstein 1567759
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 4-Chloromandelic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.