missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)benzonitrile, 98+%

Product Code. 11469657
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469657 250 g 250g
11449657 10 g 10g
11459657 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469657 Supplier Thermo Scientific Alfa Aesar Supplier No. B22138.30
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 874-86-2 | C8H6ClN | 151.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-86-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00019761
InChI Key LOQLDQJTSMKBJU-UHFFFAOYSA-N
Synonym 4-chloromethyl benzonitrile, 4-cyanobenzyl chloride, 4-chloromethyl-benzonitrile, benzonitrile, 4-chloromethyl, ccris 5103, p-cyanobenzyl chloride, 4-chloromethyl benzenecarbonitrile, 4-cyanobenzylchloride, pubchem7377, alp.-chloro-p-tolunitrile
PubChem CID 70127
IUPAC Name 4-(chloromethyl)benzonitrile
SMILES C1=CC(=CC=C1CCl)C#N

Specifications

Melting Point 77°C to 80°C
Quantity 250 g
UN Number UN2923
Formula Weight 151.6
Percent Purity ≥98%
Chemical Name or Material 4-(Chloromethyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.