missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenyl)acetic Acid Methyl Ester, TRC

Product Code. 30349050
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30349050 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349050 Supplier Toronto Research Chemicals Supplier No. TRCC378525500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4-Chlorophenyl)acetic Acid Methyl Ester
CAS 52449-43-1
Molecular Formula C9H9ClO2
Quantity 500 mg
Synonym Methyl (4-Chlorophenyl)acetate, Methyl (p-Chlorophenyl)acetate, Methyl 2-(4-Chlorophenyl)acetate, Methyl 4-Chlorobenzeneacetate, (p-Chlorophenyl)acetic Acid Methyl Ester, 4-Chlorobenzeneacetic Acid Methyl Ester
InChI Formula InChI=1S/C9H9ClO2/c1-12-9(11)6-7-2-4-8(10)5-3-7/h2-5H,6H2,1H3
SMILES COC(=O)Cc1ccc(Cl)cc1
IUPAC Name methyl 2-(4-chlorophenyl)acetate
Molecular Weight (g/mol) 184.62
Formula Weight 184.0291
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.