missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl dichlorophosphate, 98+%, Thermo Scientific™

Product Code. 10327661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10327661 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10327661 Supplier Thermo Scientific Acros Supplier No. 216100050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 772-79-2
CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Density 1.5080g/mL
Boiling Point 114.0°C to 116.0°C (1.0 mmHg)
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C6H4Cl3O2P
Linear Formula Cl2P(O)OC6H4Cl
Refractive Index 1.5380 to 1.5400
MDL Number MFCD00009705
Quantity 5 g
Specific Gravity 1.508
Synonym 4-chlorophenyl phosphorodichloridate, p-chlorophenyl dichlorophosphate, 4-chlorophenyl dichlorophosphate, 4-chlorophenylphosphorodichloridate, phosphorodichloridic acid, 4-chlorophenyl ester, 4-chlorophenylphosphoryl dichloride, acmc-1bjgo, cczmqygsxwzfki-uhfffaoysa, 4-chlorophenyl dichlorophosphinate, 4-chlorophenyl dichloridophosphate #
InChI Key CCZMQYGSXWZFKI-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl
IUPAC Name 1-chloro-4-dichlorophosphoryloxybenzene
Molecular Weight (g/mol) 245.43
PubChem CID 69879
Formula Weight 245.43
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Chlorophenyl dichlorophosphate, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chlorophenyl dichlorophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.