missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl Isocyanate, TRC

Product Code. 30350124
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350124 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350124 Supplier Toronto Research Chemicals Supplier No. TRCC377595100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chlorophenyl Isocyanate
CAS 104-12-1
Molecular Formula C7H4ClNO
Quantity 100 g
Synonym 1-Chloro-4-isocyanatobenzene, 1-Isocyanato-4-chlorobenzene, 4-Chloro-1-isocyanatobenzene, 4-Chloroisocyanatobenzene, NSC 76589, p-Chlorophenyl Isocyanate
InChI Formula InChI=1S/C7H4ClNO/c8-6-1-3-7(4-2-6)9-5-10/h1-4H
SMILES Clc1ccc(cc1)N=C=O
IUPAC Name 1-chloro-4-isocyanatobenzene
Molecular Weight (g/mol) 153.57
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.