missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl Phenyl Sulfone, TRC

Product Code. 17903365
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17903365 1 g 1g
17902955 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17903365 Supplier Toronto Research Chemicals Supplier No. TRCC3750151G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-00-2
Molecular Formula C12 H9 Cl O2 S
Molecular Weight (g/mol) 252.72
Synonym 4-Chlorodiphenyl Sulfone, 4-Chlorophenyl Phenyl Sulfone, NSC 404335, R 242, Sulfenon, Sulphenone, p-Chlorodiphenyl Sulfone, p-Chlorophenyl Phenyl Sulfone, p-Chlorophenyl Phenyl Sulfone, 1-Chloro-4-(phenylsulfonyl)benzene
IUPAC Name 1-(benzenesulfonyl)-4-chlorobenzene
SMILES Clc1ccc(cc1)S(=O)(=O)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H9ClO2S/c13-10-6-8-12(9-7-10)16(14,15)11-4-2-1-3-5-11/h1-9H
Formula Weight 252.0012
Chemical Name or Material 4-Chlorophenyl phenyl sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.