missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetylene, 98%

Product Code. 15477937
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477937 5 g 5g
15467937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477937 Supplier Thermo Scientific Alfa Aesar Supplier No. H51058.06
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 873-73-4 | C8H5Cl | 136.578 g/mol

4-Chlorophenylacetylene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenylacetylene is used as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in acetone, chloroform, dichloromethane, DMF, DMSO, ethanol, ethyl acetate, hexane, methanol,THF and toluene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-73-4
Molecular Formula C8H5Cl
Molecular Weight (g/mol) 136.578
MDL Number MFCD00191917
InChI Key LFZJRTMTKGYJRS-UHFFFAOYSA-N
Synonym 4-chlorophenylacetylene, benzene, 1-chloro-4-ethynyl, 4'-chlorophenyl acetylene, 4-chlorophenyl acetylene, 1-chloro-4-ethynyl-benzene, p-ethynylchlorobenzene, p-chlorophenylacetylene, p-chlorophenyl acetylene, 1-chlor-4-ethinylbenzol, 4-chlorophenylethyne
PubChem CID 70118
IUPAC Name 1-chloro-4-ethynylbenzene
SMILES C#CC1=CC=C(C=C1)Cl

Specifications

Melting Point 43°C to 47°C
Quantity 5 g
UN Number UN1325
Boiling Point 179°C
Solubility Information Insoluble in water. Soluble in acetone,chloroform,dichloromethane,DMF,DMSO,ethanol,ethyl acetate,hexane,methanol,THF and toluene.
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material 4-Chlorophenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.