missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenylsulfonyl)acetonitrile, 98+%, Thermo Scientific™

Product Code. 11451858
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451858 1 g 1g
11461858 5 g 5g
11471858 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451858 Supplier Thermo Scientific Alfa Aesar Supplier No. A14510.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(4-Chlorophenylsulfonyl)acetonitrile react with trimethoxymethyl-benzene, and gives 2-(4-chloro-benzenesulfonyl)-3-methoxy-3-phenyl-acrylonitrile.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1851-09-8
Molecular Formula C8H6ClNO2S
Molecular Weight (g/mol) 215.651
MDL Number MFCD00045626
InChI Key HAQGVGPNKGGSMK-UHFFFAOYSA-N
Synonym 4-chlorophenylsulfonylacetonitrile, 2-4-chlorophenyl sulfonyl acetonitrile, 4-chlorophenyl cyanomethyl sulfone, 2-4-chlorobenzenesulfonyl acetonitrile, 4-chlorobenzenesulphonylacetonitrile, 4-chlorobenzenesulfonyl acetonitrile, 4-chlorobenzenesulphonyl acetonitrile, 4-chlorophenyl sulfonyl acetonitrile, 2-4-chlorophenyl sulfonylacetonitrile, 4-chlorophenylsulfonyl acetonitrile
PubChem CID 735829
IUPAC Name 2-(4-chlorophenyl)sulfonylacetonitrile
SMILES C1=CC(=CC=C1S(=O)(=O)CC#N)Cl

Specifications

Melting Point 169°C to 173°C
Quantity 1 g
UN Number UN3439
Beilstein 1819678
Solubility Information Insoluble in water.
Formula Weight 215.66
Percent Purity ≥98%
Chemical Name or Material (4-Chlorophenylsulfonyl)acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.