Learn More
4-Cyano-3-trifluoromethylphenylisothiocyanate, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Cyano-3-trifluoromethylphenylisothiocyanate |
| CAS | 143782-23-4 |
| Molecular Formula | C9H3F3N2S |
| Quantity | 10 g |
| Synonym | 4-Isothiocyanato-2-trifluoromethylbenzonitrile |
| InChI Formula | InChI=1S/C9H3F3N2S/c10-9(11,12)8-3-7(14-5-15)2-1-6(8)4-13/h1-3H |
| SMILES | FC(F)(F)c1cc(ccc1C#N)N=C=S |
| IUPAC Name | 4-isothiocyanato-2-(trifluoromethyl)benzonitrile |
| Molecular Weight (g/mol) | 228.19 |