missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzyl alcohol, 97%

Product Code. 15400608
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400608 1 g 1g
15420608 250 mg 250mg
15410608 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400608 Supplier Thermo Scientific Alfa Aesar Supplier No. H54283.03
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 874-89-5 | C8H7NO | 133.15 g/mol

4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-89-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00870633
InChI Key XAASLEJRGFPHEV-UHFFFAOYSA-N
Synonym 4-hydroxymethyl benzonitrile, 4-cyanobenzyl alcohol, p-cyanobenzyl alcohol, ccris 5116, benzonitrile, 4-hydroxymethyl, 4-cyanobenzenemethanol, 4-cyanophenyl methanol, 4-hydroxymethyl-benzonitrile, 4 hydroxymethyl-benzonitrile
PubChem CID 160549
IUPAC Name 4-(hydroxymethyl)benzonitrile
SMILES OCC1=CC=C(C=C1)C#N

Specifications

Melting Point 32°C to 36°C
Quantity 1 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.