missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopiperidine, 96%

Product Code. 11352849
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352849 25 g 25g
11342849 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352849 Supplier Thermo Scientific Alfa Aesar Supplier No. H27722.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4395-98-6 | C6H10N2 | 110.16 g/mol

4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4395-98-6
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD05022468
InChI Key FSDNTQSJGHSJBG-UHFFFAOYSA-N
Synonym 4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349
PubChem CID 138223
IUPAC Name piperidine-4-carbonitrile
SMILES C1CNCCC1C#N

Specifications

Density 0.98
Boiling Point 226°C
Flash Point 91°C (196°F)
Odor Amine-like
Refractive Index 1.4755
Quantity 25 g
UN Number UN3276
Solubility Information Soluble in chloroform.
Formula Weight 110.16
Percent Purity 96%
Chemical Name or Material 4-Cyanopiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.