missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanostyrene, 97%, stab. with 0.05% 4-tert-butyl catechol

Product Code. 11495837
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495837 25 g 25g
11475837 1 g 1g
11485837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495837 Supplier Thermo Scientific Alfa Aesar Supplier No. B24395.14
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3435-51-6 | C9H7N | 129.16 g/mol

4-Cyanostyrene is used as an intermediate in organic synthesis. It is used in the preparation of poly(4-cyanostyrene) polymer , which is a macromolecular device having nanoscale order and finds a wide range of potential applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanostyrene is used as an intermediate in organic synthesis. It is used in the preparation of poly(4-cyanostyrene) polymer , which is a macromolecular device having nanoscale order and finds a wide range of potential applications.

Solubility
Immiscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with polymerizing initiators, polymerization catalysts and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3435-51-6
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00080445
InChI Key SNTUCKQYWGHZPK-UHFFFAOYSA-N
PubChem CID 76967
IUPAC Name 4-ethenylbenzonitrile
SMILES C=CC1=CC=C(C=C1)C#N

Specifications

Boiling Point 92°C to 93°C (3 mmHg)
Refractive Index 1.5775
Quantity 25 g
UN Number UN3276
Synonym 4-cyanostyrene, 4-vinylbenzonitrile, benzonitrile, 4-ethenyl, p-cyanostyrene, 4-vinyl-benzonitrile, benzonitrile, p-vinyl, p-vinylbenzonitrile, acmc-20amk5, benzonitrile,4-ethenyl, 4-vinylbenzenecarbonitrile
Solubility Information Immiscible with water.
Formula Weight 129.16
Percent Purity 97%
Chemical Name or Material 4-Cyanostyrene, stabilized with 0.05% 4-tert-butyl catechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.