missing translation for 'onlineSavingsMsg'
Få mere at vide

4-desamino Dapsone, TRC

Artikelnummer. 17984725
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
17984725 250 mg 250mg
17954985 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17984725 Leverandør Toronto Research Chemicals Leverandørnr. TRCP336790250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 7019-01-4
Molecular Formula C12 H11 N O2 S
Molecular Weight (g/mol) 233.29
Synonym 4-Phenylsulfonylaniline, 4-desamino Dapsone, 4-(Phenylsulfonyl)benzenamine, p-(Phenylsulfonyl)aniline, 4-(Benzenesulfonyl)aniline, 4-(Benzenesulphonyl)phenylamine, 4-(Phenylsulfonyl)benzenamine, 4-Aminodiphenyl Sulfone, 4-Aminophenyl Phenyl Sulfone, 4-Phenylsulfonylaniline, Phenyl p-Aminophenyl Sulfone, [4-(Phenylsulfonyl)phenyl]amine, p-Aminodiphenyl Sulfone, p-Aminophenyl Phenyl Sulfone, p-Phenylsulfonylaniline;
IUPAC Name 4-(benzenesulfonyl)aniline
SMILES Nc1ccc(cc1)S(=O)(=O)c2ccccc2

Tekniske data

Quantity 250 mg
InChI Formula InChI=1S/C12H11NO2S/c13-10-6-8-12(9-7-10)16(14,15)11-4-2-1-3-5-11/h1-9H,13H2
Formula Weight 233.051
Chemical Name or Material 4-Phenylsulfonylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.