missing translation for 'onlineSavingsMsg'
Learn More

4-Desmethoxypropoxyl-4-chloro Rabeprazole, TRC

Product Code. 30133253
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133253 25 mg 25mg
30007118 2.5 mg 2.5mg
30006603 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133253 Supplier Toronto Research Chemicals Supplier No. TRCD29060025MG
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168167-42-8
Molecular Formula C14 H12 Cl N3 O S
Molecular Weight (g/mol) 305.78
Synonym 2-[(RS)-[(4-Chloro-3-methylpyridin-2-yl)methyl]sulphinyl]-1H-benzimidazole, Lansoprazole Imp. F (EP)
IUPAC Name 2-[(4-chloro-3-methylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
SMILES Cc1c(Cl)ccnc1CS(=O)c2nc3ccccc3[nH]2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H12ClN3OS/c1-9-10(15)6-7-16-13(9)8-20(19)14-17-11-4-2-3-5-12(11)18-14/h2-7H,8H2,1H3,(H,17,18)
Formula Weight 305.039
Chemical Name or Material 2-[(RS)-[(4-Chloro-3-methylpyridin-2-yl)methyl]sulphinyl]-1H-benzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.