missing translation for 'onlineSavingsMsg'
Learn More

4-Desmethyl-2-methyl Celecoxib, TRC

Product Code. 30135892
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135892 250 mg 250mg
18043740 25 mg 25mg
18012540 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135892 Supplier Toronto Research Chemicals Supplier No. TRCD291815250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170569-99-0
Molecular Formula C17 H14 F3 N3 O2 S
Molecular Weight (g/mol) 381.37
Synonym Benzenesulfonamide, 4-[5-(2-methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-, 4-[5-(2-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide, 4-Desmethyl-2-methyl Celecoxib, o-Celecoxib, 4-[5-(o-Tolyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide
IUPAC Name 4-[5-(2-methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
SMILES Cc1ccccc1c2cc(nn2c3ccc(cc3)S(=O)(=O)N)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H14F3N3O2S/c1-11-4-2-3-5-14(11)15-10-16(17(18,19)20)22-23(15)12-6-8-13(9-7-12)26(21,24)25/h2-10H,1H3,(H2,21,24,25)
Formula Weight 381.0759
Percent Purity >95%
Chemical Name or Material 4-Desmethyl-2-methyl Celecoxib
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.