missing translation for 'onlineSavingsMsg'
Learn More

4-Desmethyl-3-methyl Celecoxib, TRC

Product Code. 30012822
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012822 25 mg 25mg
30010808 10 mg 10mg
30008543 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012822 Supplier Toronto Research Chemicals Supplier No. TRCD29304025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170570-01-1
Molecular Formula C17 H14 F3 N3 O2 S
Molecular Weight (g/mol) 381.37
Synonym 4-[5-(3-Methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide, Celecoxib Imp. A (Pharmeuropa)
IUPAC Name 4-[5-(3-methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
SMILES Cc1cccc(c1)c2cc(nn2c3ccc(cc3)S(=O)(=O)N)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H14F3N3O2S/c1-11-3-2-4-12(9-11)15-10-16(17(18,19)20)22-23(15)13-5-7-14(8-6-13)26(21,24)25/h2-10H,1H3,(H2,21,24,25)
Formula Weight 381.0759
Percent Purity >95%
Chemical Name or Material 4-[5-(3-methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.