missing translation for 'onlineSavingsMsg'
Learn More

4-Desmethyl-4-ethylimidazolyl Nilotinib, TRC

Product Code. 17993915
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993915 5 mg 5mg
17964395 100 mg 100mg
17950405 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17993915 Supplier Toronto Research Chemicals Supplier No. TRCD2912105MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2119583-24-1
Molecular Formula C29H24F3N7O
Molecular Weight (g/mol) 543.54
Synonym Benzamide, N-[3-(4-ethyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-, N-[3-(4-Ethyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzamide, Nilotinib ethyl homolog
SMILES O=C(NC1=CC(C(F)(F)F)=CC(N2C=C(CC)N=C2)=C1)C3=CC=C(C)C(NC4=NC(C5=CN=CC=C5)=CC=N4)=C3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C29H24F3N7O/c1-3-22-16-39(17-35-22)24-13-21(29(30,31)32)12-23(14-24)36-27(40)19-7-6-18(2)26(11-19)38-28-34-10-8-25(37-28)20-5-4-9-33-15-20/h4-17H,3H2,1-2H3,(H,36,40)(H,34,37,38)
Chemical Name or Material 4-Desmethyl-4-ethylimidazolyl Nilotinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.