missing translation for 'onlineSavingsMsg'
Learn More

4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester, 96%

Product Code. 15425326
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425326 250 mg 250mg
15415326 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425326 Supplier Thermo Scientific Alfa Aesar Supplier No. H59880.MD
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1628442-55-6 | C13H16BF3O3 | 288.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1628442-55-6
Molecular Formula C13H16BF3O3
Molecular Weight (g/mol) 288.073
MDL Number MFCD22419269
InChI Key LLJRSXMGPKHUQX-UHFFFAOYSA-N
Synonym 2-4-difluoromethoxy-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester
PubChem CID 79661309
IUPAC Name 2-[4-(difluoromethoxy)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)OC(F)F)F

Specifications

Quantity 250 mg
Formula Weight 288.07
Percent Purity 96%
Chemical Name or Material 4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.