missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino-4'-methylazobenzene, TRC

Product Code. 30179891
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179891 500 mg 500mg
30179839 50 mg 50mg
30179952 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179891 Supplier Toronto Research Chemicals Supplier No. TRCD826565500MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3010-57-9
Molecular Formula C15H17N3
Molecular Weight (g/mol) 239.32
SMILES CN(C)c1ccc(cc1)N=Nc2ccc(C)cc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H17N3/c1-12-4-6-13(7-5-12)16-17-14-8-10-15(11-9-14)18(2)3/h4-11H,1-3H3
Formula Weight 239.1422
Chemical Name or Material 4-Dimethylamino-4'-methylazobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.