missing translation for 'onlineSavingsMsg'
Learn More

4-Dodecylphenol (Mixture Of Isomers), TRC

Product Code. 30009878
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009878 5 g 5g
30005064 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009878 Supplier Toronto Research Chemicals Supplier No. TRCD0781055G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210555-94-5
Molecular Formula C18H30O
Molecular Weight (g/mol) 262.44
IUPAC Name 4-(3,5,7-trimethylnonyl)phenol
SMILES CCC(C)CC(C)CC(C)CCc1ccc(O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C18H30O/c1-5-14(2)12-16(4)13-15(3)6-7-17-8-10-18(19)11-9-17/h8-11,14-16,19H,5-7,12-13H2,1-4H3
Formula Weight 262.2297
Chemical Name or Material 4-Dodecylphenol (Mixture Of Isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.