missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenylboronic acid, 98%, Thermo Scientific™

Product Code. 10773013 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10773013 5 g Glass bottle 5g
10160172 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10773013 Supplier Thermo Scientific Acros Supplier No. 359310050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22237-13-4
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.983
MDL Number MFCD00674028
InChI Key WRQNDLDUNQMTCL-UHFFFAOYSA-N
Synonym 4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
PubChem CID 2734351
IUPAC Name (4-ethoxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OCC)(O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 165.98
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxyphenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.