missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-1,3-oxazolidin-2-one, TRC

Product Code. 30377435
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377435 500 mg
30378658 50 mg
30377970 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377435 Supplier Toronto Research Chemicals Supplier No. TRCE937813500MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16112-60-0
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
IUPAC Name 4-ethyl-1,3-oxazolidin-2-one
SMILES CCC1COC(=O)N1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H9NO2/c1-2-4-3-8-5(7)6-4/h4H,2-3H2,1H3,(H,6,7)
Formula Weight 115.0633
Chemical Name or Material 4-Ethyl-1,3-oxazolidin-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.