Learn More
4-Ethyl-3-nitrobenzoic Acid, TRC
Product Code. 30373934
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373934 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30373934 Supplier Toronto Research Chemicals Supplier No. TRCE9372455G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Ethyl-3-nitrobenzoic Acid
CAS 103440-95-5
Molecular Formula C9 H9 N O4
Quantity 5 g
InChI Formula InChI=1S/C9H9NO4/c1-2-6-3-4-7(9(11)12)5-8(6)10(13)14/h3-5H,2H2,1H3,(H,11,12)
SMILES CCc1ccc(cc1[N+](=O)[O-])C(=O)O
IUPAC Name 4-ethyl-3-nitrobenzoic acid
Molecular Weight (g/mol) 195.172
Formula Weight 195.053