missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzaldehyde, 97%

Product Code. 11365319
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365319 25 g 25g
11375319 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365319 Supplier Thermo Scientific Alfa Aesar Supplier No. B20645.14
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4748-78-1 | C9H10O | 134.178 g/mol

It is used as a perfuming agent in cosmetics industry. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a perfuming agent in cosmetics industry. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4748-78-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00006956
InChI Key QNGNSVIICDLXHT-UHFFFAOYSA-N
Synonym p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p
PubChem CID 20861
IUPAC Name 4-ethylbenzaldehyde
SMILES CCC1=CC=C(C=C1)C=O

Specifications

Density 0.979
Boiling Point 220°C to 221°C
Flash Point 92°C (197°F)
Odor Characteristic
Refractive Index 1.539
Quantity 25 g
Beilstein 636027
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Ethylbenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.