missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylbenzoic acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-Ethylbenzoic acid |
| CAS | 619-64-7 |
| Molecular Formula | C9H10O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C9H10O2/c1-2-7-3-5-8(6-4-7)9(10)11/h3-6H,2H2,1H3,(H,10,11) |
| SMILES | CCc1ccc(cc1)C(=O)O |
| Molecular Weight (g/mol) | 150.18 |
| Formula Weight | 150.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction