missing translation for 'onlineSavingsMsg'
Learn More

(4-Ethylphenyl)boronic acid, 98%, BLD Pharm™

Product Code. 31106462
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
10 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31106462 10 g
31099465 500 g
31092694 100 g
31095150 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31106462 Supplier BLD Pharm Supplier No. BD1066310g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 63139-21-9
Molecular Formula C8H11BO2
MDL Number MFCD00859377
Quantity 10 g
SMILES CCC1=CC=C(B(O)O)C=C1
Molecular Weight (g/mol) 149.98
Percent Purity 98%
Chemical Name or Material (4-Ethylphenyl)boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.