missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Product Code. 10311390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311390 50 mL AcroSeal™ Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10311390 Supplier Thermo Scientific Acros Supplier No. 431890500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 22873-28-5 | C8H9BrMg | 209.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22873-28-5, 109-99-9
CAS Min % 8.82, 86.78
CAS Max % 13.22, 91.18
Color Yellow to Brown
Density 0.9500g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C8H9BrMg
Linear Formula CH3CH2C6H4MgBr
MDL Number MFCD01311467
Quantity 50 mL
Specific Gravity 0.95
Synonym 4-ethylphenylmagnesium bromide, 4-ethylphenyl magnesium bromide, vlouakpiqasfds-uhfffaoysa-m, 4-ethylphenylmagnesium bromide, 0.5m in 2-methf, 4-ethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 4-ethylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key QSGSHZBZGXYVOQ-UHFFFAOYSA-M
SMILES [Mg++].[Br-].CCC1=CC=[C-]C=C1
IUPAC Name magnesium;ethylbenzene;bromide
Molecular Weight (g/mol) 209.37
PubChem CID 11241259
Formula Weight 209.37
Physical Form Solution
Chemical Name or Material 4-Ethylphenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Ethylphenylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.