missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2,3-dimethylaniline, 97%

Product Code. 11315175
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315175 1 g 1g
15404157 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11315175 Supplier Thermo Scientific Alfa Aesar Supplier No. H31560.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1737-68-4 | C8H10FN | 139.173 g/mol

4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1737-68-4
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD04972870
InChI Key LKMJLYCMLAPNDB-UHFFFAOYSA-N
Synonym 3-amino-6-fluoro-1,2-dimethylbenzene, 4-fluoro-2,3-dimethyl aniline, 2,3-dimethyl-4-fluoroaniline, 4-fluoro-o-xylidine, benzenamine, 4-fluoro-2,3-dimethyl, 4-fluoro-2,3-dimethyl-phenylamine, p-fluordimethylanilin, intermediates-zcf02293, 4-fluoranyl-2,3-dimethyl-aniline, 4-fluoro-2,3-dimethyl-benzenamine
PubChem CID 17770833
IUPAC Name 4-fluoro-2,3-dimethylaniline
SMILES CC1=C(C=CC(=C1C)F)N

Specifications

Melting Point 42°C to 46°C
Quantity 1 g
UN Number UN2811
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2,3-dimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.