missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylphenylboronic acid, TRC

Product Code. 30405173
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30405173

Brand: Toronto Research Chemicals TRCF596520100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluoro-2-methylphenylboronic acid
CAS 139911-29-8
Molecular Formula C7 H8 B F O2
Quantity 100 mg
Synonym B-(4-Fluoro-2-methylphenyl)boronic Acid, (2-Methyl-4-fluorophenyl)boronic Acid
InChI Formula InChI=1S/C7H8BFO2/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4,10-11H,1H3
SMILES Cc1cc(F)ccc1B(O)O
IUPAC Name (4-fluoro-2-methylphenyl)boronic acid
Molecular Weight (g/mol) 153.947
Formula Weight 154.06
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.